1-Bromo-3,4,5-trifluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 5-Bromo-1,2,3-trifluorobenzene

001801-25G 001801-100G
FLUO001801-25GEA 0 GBP
FLUO001801-25G FLUO001801-100G
1-Bromo-3,4,5-trifluorobenzene
1-Bromo-3,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 140
Melting Pt: –20 °C
Density: 1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00070738
CAS Number: 138526-69-9
EINECS: 418-480-9
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR