1-Bromo-2,6-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Bromo-1,3-difluorobenzene

001117-25G 001117-10G 001117-100G
FLUO001117-25GEA 0 GBP
FLUO001117-25G FLUO001117-10G FLUO001117-100G
1-Bromo-2,6-difluorobenzene
1-Bromo-2,6-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 61
Density: 1.71 g/cm³ (20 °C)
MDL Number: MFCD00009894
CAS Number: 64248-56-2
EINECS: 264-750-2
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR