2-Bromo-1,4-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,5-Difluorobromobenzene, 1-Bromo-2,5-difluorobenzene

001800-25G 001800-100G
FLUO001800-25GEA 0 GBP
FLUO001800-25G FLUO001800-100G
2-Bromo-1,4-difluorobenzene
2-Bromo-1,4-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 158 °C (1013 hPa)
Melting Pt: –32 °C
Density: 1,7414 g/cm³ (20 °C)
MDL Number: MFCD00000345
CAS Number: 399-94-0
EINECS: 206-920-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR