1-Bromo-2,4-difluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
001462-100G
001462-250G
001462-1KG
FLUO001462-100GEA
0
GBP
FLUO001462-100G
FLUO001462-250G
FLUO001462-1KG
1-Bromo-2,4-difluorobenzene
1-Bromo-2,4-difluorobenzene
Formula:
C₆H₃BrF₂ Boiling Pt: 145 °C (1013 hPa) Melting Pt: –4 °C Density: 1.69 g/cm³ (20 °C) Flash Pt: 51 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000330 CAS Number: 348-57-2 EINECS: 206-479-4 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...