1-Bromo-2,4,5-trifluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001622-250G 001622-100G 001622-25G
FLUO001622-250GEA 0 GBP
FLUO001622-250G FLUO001622-100G FLUO001622-25G
1-Bromo-2,4,5-trifluorobenzene
1-Bromo-2,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
Boiling Pt: 144 °C (1013 hPa)
Melting Pt: –19 °C
Density: 1.8022 g/cm³ (20 °C)
MDL Number: MFCD00000306
CAS Number: 327-52-6
EINECS: 206-318-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR