1-Bromo-2,3-difluorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2,3-Difluorobromobenzene

001797-1G 001797-5G 001797-25G
FLUO001797-1GEA 0 GBP
FLUO001797-1G FLUO001797-5G FLUO001797-25G
1-Bromo-2,3-difluorobenzene
1-Bromo-2,3-difluorobenzene
Formula: C₆H₃BrF₂
MW: 192.99 g/mol
Boiling Pt: 234 °C (765 mmHg)
Density: 1.72 g/cm³ (20 °C)
MDL Number: MFCD00061136
CAS Number: 38573-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR