1-Bromo-2,3,5,6-tetrafluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2,3,5,6-Tetrafluorobromobenzene, 3-Bromo-1,2,4,5-tetrafluorobenzene
010257-1G
010257-5G
010257-25G
FLUO010257-1GEA
0
GBP
FLUO010257-1G
FLUO010257-5G
FLUO010257-25G
1-Bromo-2,3,5,6-tetrafluorobenzene
1-Bromo-2,3,5,6-tetrafluorobenzene
Formula:
C₆HBrF₄ MW: 228.97 g/mol Boiling Pt: 143 Density: 1.88 g/cm³ (20 °C) |
MDL Number:
MFCD00000308 CAS Number: 1559-88-2 EINECS: 216-327-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...