2,5-Dibromo-1-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1,4-Dibromo-2-fluorobenzene
004652-25G
004652-100G
FLUO004652-25GEA
0
GBP
FLUO004652-25G
FLUO004652-100G
2,5-Dibromo-1-fluorobenzene
2,5-Dibromo-1-fluorobenzene
Formula:
C₆H₃Br₂F Boiling Pt: 216 °C (1013 hPa) Melting Pt: 35 °C |
MDL Number:
MFCD00010603 CAS Number: 1435-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...