4-Bromo-3-chloroaniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003693-25G 003693.5G 003693-1G 003693-100G
FLUO003693-25GEA 0 GBP
FLUO003693-25G FLUO003693.5G FLUO003693-1G FLUO003693-100G
4-Bromo-3-chloroaniline
4-Bromo-3-chloroaniline
Formula: C₆H₅BrClN
MW: 206.47 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 67…68 °C
MDL Number: MFCD00041311
CAS Number: 21402-26-6
EINECS: 244-370-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR