4-Bromo-2-chloroaniline
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003692-25G
003692-100G
003692-10G
FLUO003692-25GEA
0
GBP
FLUO003692-25G
FLUO003692-100G
FLUO003692-10G
4-Bromo-2-chloroaniline
4-Bromo-2-chloroaniline
Formula:
C₆H₅BrClN Boiling Pt: 264 °C (1013 hPa) Melting Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007660 CAS Number: 38762-41-3 EINECS: 254-118-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...