1-Bromo-3-chloro-4-fluorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-Bromo-2-chloro-1-fluorobenzene, 4-Bromo-2-chlorofluorobenzene
002876-100G
002876-250G
FLUO002876-100GEA
0
GBP
FLUO002876-100G
FLUO002876-250G
1-Bromo-3-chloro-4-fluorobenzene
1-Bromo-3-chloro-4-fluorobenzene
Formula:
C₆H₃BrClF Boiling Pt: 92 °C (35 mmHg) Density: 1.72 g/cm³ (20 °C) |
MDL Number:
MFCD00051794 CAS Number: 60811-21-4 EINECS: 405-580-2 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...