1-Bromo-3-chlorobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromochlorobenzene
001104-100G
001104-500G
FLUO001104-100GEA
0
GBP
FLUO001104-100G
FLUO001104-500G
1-Bromo-3-chlorobenzene
1-Bromo-3-chlorobenzene
Formula:
C₆H₄BrCl MW: 191.45 g/mol Boiling Pt: 196 °C (1013 hPa) Melting Pt: –21,2 °C Density: 1,63 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000568 CAS Number: 108-37-2 EINECS: 203-575-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...