1-Bromo-2-chlorobenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001103-100G 001103-250G
FLUO001103-100GEA 0 GBP
FLUO001103-100G FLUO001103-250G
1-Bromo-2-chlorobenzene
1-Bromo-2-chlorobenzene
Formula: C₆H₄BrCl
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: –12,1 °C
Density: 1,65 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000532
CAS Number: 694-80-4
EINECS: 211-775-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR