5-Bromoanthranilic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Amino-5-bromobenzoic acid

018242-10G 018242-100G 018242-25G
FLUO018242-10GEA 0 GBP
FLUO018242-10G FLUO018242-100G FLUO018242-25G
5-Bromoanthranilic acid
5-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 218…219 °C
Storage Temperature: Ambient
MDL Number: MFCD00007823
CAS Number: 5794-88-7
EINECS: 227-338-3
UN: 2811
ADR: 6.1,III
Merck Index: 13,01389

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR