2-Bromobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

003019-100G 003019-500G
FLUO003019-100GEA 0 GBP
FLUO003019-100G FLUO003019-500G
2-Bromobenzoic acid
2-Bromobenzoic acid
Formula: C₇H₅BrO₂
Boiling Pt: 296 °C (1013 hPa)
Melting Pt: 144…147 °C
Density: 1,929 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002402
CAS Number: 88-65-3
EINECS: 201-848-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR