4-Bromobenzonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
p-Bromobenzonitrile
002988-25G
002988-100G
FLUO002988-25GEA
0
GBP
FLUO002988-25G
FLUO002988-100G
4-Bromobenzonitrile
4-Bromobenzonitrile
Formula:
C₇H₄BrN Boiling Pt: 236 °C (1013 hPa) Melting Pt: ∼90 °C Density: 1.562 g/cm³ (20 °C) Flash Pt: 130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001811 CAS Number: 623-00-7 EINECS: 210-764-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...