1,3-Dibromobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
001159-100G
001159-500G
001159-25G
FLUO001159-100GEA
0
GBP
FLUO001159-100G
FLUO001159-500G
FLUO001159-25G
1,3-Dibromobenzene
1,3-Dibromobenzene
Formula:
C₆H₄Br₂ Boiling Pt: 218 °C (1013 hPa) Melting Pt: –7 °C Density: 1,952 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000078 CAS Number: 108-36-1 EINECS: 203-574-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...