1,2-Dibromobenzene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001158-100G
001158-25G
001158-250G
FLUO001158-100GEA
0
GBP
FLUO001158-100G
FLUO001158-25G
FLUO001158-250G
1,2-Dibromobenzene
1,2-Dibromobenzene
Formula:
C₆H₄Br₂ Boiling Pt: 224 °C (1013 hPa) Density: 1.956 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000057 CAS Number: 583-53-9 EINECS: 209-507-3 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...