9,10-Diphenylanthracene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002922-5G 002922-25G
FLUO002922-5GEA 0 GBP
FLUO002922-5G FLUO002922-25G
9,10-Diphenylanthracene
9,10-Diphenylanthracene
Formula: C₂₆H₁₈
Boiling Pt: 485 °C (1013 hPa)
Melting Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00001253
CAS Number: 1499-10-1
EINECS: 216-105-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR