1H,1H,2H-Perfluoro-1-decene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
003394-25G
003394-100G
FLUO003394-25GEA
0
GBP
FLUO003394-25G
FLUO003394-100G
1H,1H,2H-Perfluoro-1-decene
1H,1H,2H-Perfluoro-1-decene
Formula:
C₁₀H₃F₁₇ MW: 446.11 g/mol Boiling Pt: 145…150 °C (1013 hPa) Density: 1,65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00039246 CAS Number: 21652-58-4 EINECS: 244-503-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...