1H,1H,2H-Perfluoro-1-octene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003395-25G 003395-100G 003395-5G
FLUO003395-25GEA 0 GBP
FLUO003395-25G FLUO003395-100G FLUO003395-5G
1H,1H,2H-Perfluoro-1-octene
1H,1H,2H-Perfluoro-1-octene
Formula: C₈H₃F₁₃
Boiling Pt: 100 °C (1013 hPa)
Density: 1.56 g/cm³ (20 °C)
MDL Number: MFCD00039249
CAS Number: 25291-17-2
EINECS: 246-791-8
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR