1H,1H,2H-Perfluoro-1-hexene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H,1H,2H-Nonafluorohex-1-ene
007057-25G
007057-100G
FLUO007057-25GEA
0
GBP
FLUO007057-25G
FLUO007057-100G
1H,1H,2H-Perfluoro-1-hexene
1H,1H,2H-Perfluoro-1-hexene
Formula:
C₆H₃F₉ MW: 246.08 g/mol Boiling Pt: 59…60 °C (1013 hPa) Density: 1.41 g/cm³ (20 °C) |
MDL Number:
MFCD00042338 CAS Number: 19430-93-4 EINECS: 243-053-7 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...