Trichloroethylene ≥99.5%, Electronic Grade
Supplier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Purity: 99.5+%
CAS No.: 79-01-6
Molecular Formula: ClCH=CCl2
Formula Weight: 131.39
Flash Point: None
Physical Form: Liquid
Density: 1.46
Refractive Index: 1.476
Odour: Chloroform-like
Melting Point: -85°
Boiling Point: 87°
Beilstein Reference: 1736782
Merck Reference: 14,9639
UN No.: 1710
MDL No.: MFCD00000838
Formula:
ClCH=CCl₂ Boiling Pt: 87 °C Melting Pt: –85 °C Density: 1.46 Storage Temperature: Ambient |
MDL Number:
MFCD00000838 CAS Number: 79-01-6 EINECS: 201-167-4 UN: 1710 ADR: 6.1,III Merck Index: 14,9639 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...