4-Bromo-1-butene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010654-10G 010654-50G 010654-5G 010654-25G
FLUO010654-10GEA 0 GBP
FLUO010654-10G FLUO010654-50G FLUO010654-5G FLUO010654-25G
4-Bromo-1-butene
4-Bromo-1-butene
Formula: C₄H₇Br
Boiling Pt: 98 °C (1013 hPa)
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1992
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR