2-Methylpentane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 1,1-Dimethylbutane, Isohexane

MQ200

M65807-5G M65807-100G
SIALM65807-5GEA 36 GBP
SIALM65807-5G SIALM65807-100G
2-Methylpentane, Sigma-Aldrich®
2-Methylpentane
Formula: C₆H₁₄
MW: 86.18 g/mol
Boiling Pt: 62
Melting Pt: –154 °C
Density: 0.653 g/cm³ (20 °C)
Flash Pt: –33 °C
Storage Temperature: Ambient
MDL Number: MFCD00009406
CAS Number: 107-83-5
EINECS: 203-523-4
UN: 1208
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ce61b1646b637970765dc4b16a997650aeccfd1&vid=71606860b34952f73fd29b7df735dd0916b1e93c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR