1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5,5-Dimethyl-1,1,1-trifluorohexane-2,4-dione

001330-5G 001330-25G
FLUO001330-5GEA 0 GBP
FLUO001330-5G FLUO001330-25G
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione
1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione
Formula: C₈H₁₁F₃O₂
MW: 196.17 g/mol
Boiling Pt: 178 °C (1013 hPa)
Density: 1.129 g/cm³ (20 °C)
MDL Number: MFCD00040841
CAS Number: 22767-90-4
EINECS: 245-211-0
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR