6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyloctane-3,5-dione, 1,1,1,2,2,3,3-Heptafluoro-7,7-dimethyl-4,6-octanedione, HFOD
001431-5G
001431-25G
FLUO001431-5GEA
0
GBP
FLUO001431-5G
FLUO001431-25G
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
6,6,7,7,8,8,8-Heptafluoro-2,2-dimethyl-3,5-octanedione
Formula:
C₁₀H₁₁F₇O₂ Boiling Pt: 173 °C (1013 hPa) Melting Pt: 34…35 °C Density: 1.27 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000435 CAS Number: 17587-22-3 EINECS: 241-556-6 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...