2,2,6,6-Tetramethyl-3,5-heptanedione

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Dipivaloylmethane, TMHD, 2,2,6,6-Tetramethylheptane-3,5-dione

001030-25G 001030-5G
FLUO001030-25GEA 0 GBP
FLUO001030-25G FLUO001030-5G
2,2,6,6-Tetramethyl-3,5-heptanedione
2,2,6,6-Tetramethyl-3,5-heptanedione
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
Boiling Pt: 203
Melting Pt: 28…30 °C
Density: 0.88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008848
CAS Number: 1118-71-4
EINECS: 214-268-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR