Isobutyraldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-Methylpropanal, 2-Methylpropionaldehyde
MQ100
418110-100ML
418110-800ML
SIAL418110-100MLEA
134
GBP
SIAL418110-100ML
SIAL418110-800ML
Isobutyraldehyde, Sigma-Aldrich®
Isobutyraldehyde
Formula:
C₄H₈O MW: 72.11 g/mol Boiling Pt: 64 Melting Pt: –65 °C Density: 0.794 g/cm³ (20 °C) Flash Pt: –24 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006980 CAS Number: 78-84-2 EINECS: 201-149-6 UN: 2045 ADR: 3 II Merck Index: 13,05171 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...