n-Tricosane 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
421600050
421600010
421600250
ACRO421600050EA
48
GBP
ACRO421600050
ACRO421600010
ACRO421600250
n-Tricosane 98%
n-Tricosane
Formula:
H₃C(CH₂)₂₁CH₃ Boiling Pt: 380 °C (760 mmHg) Melting Pt: 47…50 °C Density: 0,7969 g/cm³ (20 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009350 CAS Number: 638-67-5 EINECS: 211-347-4 |
Specification Test Results
Appearance | White Crystals, flakes and/or chunks |
Infrared spectrum | Conforms |
Melting point | 47 to 50 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...