n-Triacontane 98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
278050010
278050050
ACRO278050010EA
41.5
GBP
ACRO278050010
ACRO278050050
n-Triacontane 98%
n-Triacontane
Formula:
H₃C(CH₂)₂₈CH₃ MW: 422.82 g/mol Boiling Pt: 455 °C (760 mmHg) Melting Pt: 65…67 °C Flash Pt: 238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009410 CAS Number: 638-68-6 EINECS: 211-349-5 |
Specification Test Results
Appearance | White Glistening flakes |
Infrared spectrum | Conforms |
Melting point | 65 to 67 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...