n-Tetracosane 99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N-tetracosane

138040250 138041000
ACRO138040250EA 37.8 GBP
ACRO138040250 ACRO138041000
n-Tetracosane 99%
n-Tetracosane
Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 391.3
Melting Pt: 49…52 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00009352
CAS Number: 646-31-1
EINECS: 211-474-5

Order Now

Specification Test Results

Appearance White Glistening flakes
Infrared spectrum Conforms
Melting point 49 to 52 °C
GC ≥98.5 %
Color scale ≤10 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR