Propargyl amine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

M-Clarity™ quality level = MQ200

P50900-5G P50900-1G P50900-25G
SIALP50900-5GEA 158 GBP
SIALP50900-5G SIALP50900-1G SIALP50900-25G
Propargyl amine, Sigma-Aldrich®
Propagyl amine
Formula: C₃H₅N
Boiling Pt: 83 °C (1013 hPa)
Density: 0.867 g/cm³ (16 °C)
MDL Number: MFCD00008198
CAS Number: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR