1-Acetylimidazole, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(1H-Imidazol-1-yl)ethanone, 1-Acetyl-1H-imidazole

M-Clarity™ quality level = MQ200

157864-100G 157864-25G
SIAL157864-100GEA 180 GBP
SIAL157864-100G SIAL157864-25G
1-Acetylimidazole, Sigma-Aldrich®
1-Acetylimidazole
Formula: C₅H₆N₂O
MW: 110.12 g/mol
MDL Number: MFCD00005287
CAS Number: 2466-76-4
EINECS: 219-577-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR