4'-Phenoxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018943-100G 018943-25G
FLUO018943-100GEA 0 GBP
FLUO018943-100G FLUO018943-25G
4'-Phenoxyacetophenone
4'-Phenoxyacetophenone
Formula: C₁₄H₁₂O₂
Boiling Pt: 119…120 °C (0,03 mmHg)
Melting Pt: 56 °C
MDL Number: MFCD00008744
CAS Number: 5031-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR