1H,1H-Heneicosafluoro-1-undecanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-Heneicosafluoro-1-undecanol, 1H,1H-Perfluoro-1-undecanol, 1H,1H-Perfluoroundecan-1-ol
007176-5G
007176-25G
FLUO007176-5GEA
0
GBP
FLUO007176-5G
FLUO007176-25G
1H,1H-Heneicosafluoro-1-undecanol
1H,1H-Heneicosafluoro-1-undecanol
Formula:
C₁₁H₃F₂₁O MW: 550.11 g/mol |
MDL Number:
MFCD00153269 CAS Number: 307-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...