3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
007055-5G
007055-25G
FLUO007055-5GEA
0
GBP
FLUO007055-5G
FLUO007055-25G
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
3,3,4,4,5,5,6,6,6-Nonafluoro-1-hexanol
Formula:
C₆H₅F₉O |
MDL Number:
MFCD00039543 CAS Number: 2043-47-2 EINECS: 218-050-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...