1H,1H-Tridecafluoro-1-heptanol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1H,1H-Perfluoro-1-heptanol, 2,2,3,3,4,4,5,5,6,6,7,7,7-Tridecafluoro-1-heptanol
007047-5G
007047-25G
FLUO007047-5GEA
0
GBP
FLUO007047-5G
FLUO007047-25G
1H,1H-Tridecafluoro-1-heptanol
1H,1H-Tridecafluoro-1-heptanol
Formula:
C₇H₃F₁₃O |
MDL Number:
MFCD00042433 CAS Number: 375-82-6 EINECS: 206-796-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...