1,3-Dibromo-1,1-difluorobutane

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

004642-5G 004642-25G
FLUO004642-5GEA 0 GBP
FLUO004642-5G FLUO004642-25G
1,3-Dibromo-1,1-difluorobutane
1,3-Dibromo-1,1-difluorobutane
Formula: C₄H₆Br₂F₂
CAS Number: 406-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR