1H,1H,9H-Hexadecafluoro-1-nonanol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003249-25G 003249-100G 003249-250G
FLUO003249-25GEA 0 GBP
FLUO003249-25G FLUO003249-100G FLUO003249-250G
1H,1H,9H-Hexadecafluoro-1-nonanol
1H,1H,9H-Hexadecafluoro-1-nonanol
Formula: C₉H₄F₁₆O
MDL Number: MFCD00039629
CAS Number: 376-18-1
EINECS: 206-806-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR