2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002304-5G
002304-25G
002304-100G
FLUO002304-5GEA
0
GBP
FLUO002304-5G
FLUO002304-25G
FLUO002304-100G
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
2,2,3,3,4,4,5,5-Octafluoro-1,6-hexanediol
Formula:
C₆H₆F₈O₂ Boiling Pt: 135…136 °C (27 mmHg) Melting Pt: 69 °C |
MDL Number:
MFCD00069087 CAS Number: 355-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...