4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001248-100G
001248-250G
001248-50G
FLUO001248-100GEA
0
GBP
FLUO001248-100G
FLUO001248-250G
FLUO001248-50G
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
4,4,4-Trifluoro-1-phenyl-1,3-butanedione
Formula:
C₁₀H₇F₃O₂ |
MDL Number:
MFCD00000425 CAS Number: 326-06-7 EINECS: 206-307-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...