Potassium(2-naphthalene)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Potassium 2-Naphthalenetrifluoroborate, (2-Naphthalene)trifluoroboric acid potassium salt, Potassium trifluoro(naphthalen-2-yl)borate, Potassium trifluoro(2-naphthalenyl)borate

TB-3256-1G TB-3256-25G TB-3256-5G
COBBTB-3256-1GEA 0 GBP
COBBTB-3256-1G COBBTB-3256-25G COBBTB-3256-5G
Potassium(2-naphthalene)trifluoroborate
Potassium (2-naphthalene)trifluoroborate
Formula: C₁₀H₇BF₃K
MDL Number: MFCD06659941
CAS Number: 668984-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR