Potassium (2-nitrophenyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-3232-1G TB-3232-25G TB-3232-5G
COBBTB-3232-1GEA 0 GBP
COBBTB-3232-1G COBBTB-3232-25G COBBTB-3232-5G
Potassium (2-nitrophenyl)trifluoroborate
Potassium (2-nitrophenyl)trifluoroborate
Formula: C₆H₄BF₃KNO₂
MDL Number: MFCD04115759
CAS Number: 850623-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR