Potassium (3-cyanophenyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Cyanophenyl)trifluoroboric acid potassium salt

TB-3211-1G TB-3211-25G TB-3211-5G
COBBTB-3211-1GEA 15.83 GBP
COBBTB-3211-1G COBBTB-3211-25G COBBTB-3211-5G
Potassium (3-cyanophenyl)trifluoroborate
Potassium (3-cyanophenyl)trifluoroborate
Formula: C₇H₄BF₃KN
MDL Number: MFCD04115744
CAS Number: 850623-46-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR