Potassium (3-methoxyphenyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Potassium trifluoro(3-methoxyphenyl)borate, (3-Methoxyphenyl)trifluoroboric acid potassium salt

TB-3204-1G TB-3204-25G TB-3204-5G
COBBTB-3204-1GEA 0 GBP
COBBTB-3204-1G COBBTB-3204-25G COBBTB-3204-5G
Potassium (3-methoxyphenyl)trifluoroborate
Potassium (3-methoxyphenyl)trifluoroborate
Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
MDL Number: MFCD04039954
CAS Number: 438553-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR