Sodium decyl sulphate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Decyl sodium sulphate, Decyl sodium sulfate, Decyl sulphuric acid salt, Decyl sulfuric acid salt

01075.25G 001075-10G 001075-25G
FLUO01075.25GEA 0 GBP
FLUO01075.25G FLUO001075-10G FLUO001075-25G
Sodium decyl sulphate
Sodium decyl sulphate
Formula: C₁₀H₂₁NaO₄S
MW: 260.33 g/mol
Melting Pt: 191,85 °C
Density: 1,207 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00041881
CAS Number: 142-87-0
EINECS: 205-568-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR