1-Tetracosanol 99+%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Tricosyl carbinol, 1-Tetracosyl alcohol, Tetracosanol, Lignoceryl alcohol, 1-Hydroxy tetracosane
258150010
258151000
ACRO258150010EA
743
GBP
ACRO258150010
ACRO258151000
1-Tetracosanol 99+%
1-Tetracosanol
Formula:
H₃C(CH₂)₂₂CH₂OH MW: 354.66 g/mol Boiling Pt: 190 Melting Pt: 75…77 °C Flash Pt: 218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042662 CAS Number: 506-51-4 EINECS: 208-043-9 |
Specification Test Results
Appearance | White Glistening powder or flakes |
Infrared spectrum | Conforms |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...