1-Hexanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Hexyl alcohol, Pentyl carbinol, 1-Hexyl alcohol, 1-Hydroxy hexane

M-Clarity™ quality level = MQ100

52828-25ML 52828-5ML
SIAL52828-25MLEA 182 GBP
SIAL52828-25ML SIAL52828-5ML
1-Hexanol, Sigma-Aldrich®
1-Hexanol
Formula: H₃C(CH₂)₄CH₂OH
MW: 102.18 g/mol
Boiling Pt: 156 °C (1013 hPa)
Melting Pt: –45 °C
Density: 0.8322 g/cm³ (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00002982
CAS Number: 111-27-3
EINECS: 203-852-3
UN: 2282
ADR: 3,III
Merck Index: 13,04715
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a0793e56caf8b4e459c31d9f36b2eec3d50a4e4&vid=5117270d1d57dd4b9cca9ee3d787375106cc1216&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR