1-Heptadecanol

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Heptadecan-1-ol, Hexadecyl carbinol, 1-Heptadecanol

L20404.06 L20404.14
L20404.06EA 47.4 GBP
L20404.06 L20404.14
1-Heptadecanol
1-Heptadecanol
Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
Boiling Pt: 308…309 °C
Melting Pt: 52…56 °C
Density: 0.967 g/cm³ (20 °C)
Flash Pt: 155 °C (311 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002822
CAS Number: 1454-85-9
EINECS: 215-932-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR